Nalgene™ HDPE, Jerry Can with Tethered Polypropylene Closure (Double Port)
Thermo Scientific™
Nalgene™ HDPE, Jerry Can with Tethered Polypropylene Closure (Double Port)
For Use With: Ideal as a reservoir in automated systems; Closure Material: Polypropylene; Color: White
Have Questions?
Catalog number 2243-9013
Price (USD)
630.00
Case of 4
Price (USD)
630.00
Case of 4
Store reagents with the Thermo Scientific™ Nalgene™ Jerrican with Tethered Polypropylene Closure, featuring a space-saving design, molded-in handle and recessed grip at the bottom to provide better control when pouring. The double-ported 2243-9013 is ideal as a reservoir in automated systems. The 53B closure can be plumbed with input/output tubing while the 38mm closure is used for refilling or emptying the reservoir.
Key Features:
  • Jerrican is ideal as a reservoir in automated systems like the PRISM™ 3700 DAN Analyzer
  • Molded-in graduations in 1 gal. and 5L increments allow for ease in dispensing
  • Molded-in handle provides a sturdy grip when carrying heavy contents
  • Integral shoulder handle and molded bottom grip make lifting comfortable
  • Wide stance and low center of gravity add stability
  • Heavy rugged design with good chemical resistance allows for a wide variety of solutions
  • Space-saving rectangular shape permits efficient storage
Ordering Information:
  • For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
  • Custom tubing sets can be designed at the Thermo Scientific Custom Tubing Set Configurator, www.thermoscientific.com/tubingconfigurator
Specifications
Capacity (English)3.5 gal.
Certifications/ComplianceUSP Class VI Compliant/FDA21 CFR 177.1520 Compliant
Closure ColorWhite
Closure MaterialPolypropylene
Closure Size53B
Closure TypeTethered
Diameter (English) Inner Neck1.5 in.
Diameter (Metric) Inner Neck38 mm
For Use With (Application)Ideal as a reservoir in automated systems
No. per Case4/Cs.
No. per Pack1 Ea.
Capacity (Metric)13 L
ColorWhite
MaterialHDPE
Product TypeJerry Can
Unit SizeCase of 4