MEM α, no nucleosides, 10 x 500 mL - FAQs
View additional product information for MEM α, no nucleosides - FAQs (22561054)
1 product FAQs found
Where can I find the osmolality for MEM Medium?
The osmolality is listed in the COA for the particular lot number of the medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.