Magnesium stearate (vegetable based), Hyqual™, GenAR™, FCC, NF, GMP, Macron Fine Chemicals™
Avantor™

Magnesium stearate (vegetable based), Hyqual™, GenAR™, FCC, NF, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets NF, FCC, USP, JP, ChP, BP and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
12 kg
Catalog number 2257-06
Price (USD)
-
Quantity:
2.5 kg
This product is GMP Manufactured and meets NF, FCC, USP, JP, ChP, BP and EP specifications.
This product is GMP Manufactured and meets NF, FCC, USP, JP, ChP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS557-04-0
Chemical Name or MaterialMagnesium stearate
ColorWhite
Melting Point130°C to 140°C
Molecular FormulaC36H70MgO4
PackagingMall STAKMOR case
SynonymStearic acid Magnesium(II) salt, Stearic acid magnesium salt, Magnesium(II) Stearate
FormPowder
Molecular Weight (g/mol)591.25
Quantity2.5 kg
Unit SizeEach