Bacto™ Casitone
Gibco™

Bacto™ Casitone

Bacto Casitone is an animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest. While this supplementRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
22591010 kg
225930500 g
Catalog number 225910
Price (MXN)
-
Quantity:
10 kg
Bacto Casitone is an animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest. While this supplement provides a nutritional profile based on casein, the specific enzymatic digest performed for this product results in a particularly high content of amino acids and peptides of varying sizes, making it a more nutritious hydrolysate. Customers have successfully used this supplement for the production of Escherichia coli-based recombinant proteins.

Specifications
For Use With (Application)Production of Escherichia coli-based recombinant proteins
Product LineBacto
Quantity10 kg
SourceAnimal Origin, Casein
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.

Frequently asked questions (FAQs)

What alternative product do you recommend for Bacto Tryptone (Cat. No. 211699, 211701, 211705)?

We have several casein digests that could serve as alternatives to Bactone Tryptone (Cat. No. 211699, 211791, 211705), including Bacto Casitone (Cat. No. 225930, 225910), Difco Tryptone Peptone (Cat. No. 21921, 211922, 211923), BiTek Tryptone (Cat. No. 251420). However, the digestion and nutritive profile of each peptone are different, therefore there may be differences in bacterial growth or supplementation ability. We recommend empirically determining the performance of alternative casein digestion in your application.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.