Stachyose hydrate, 97+%
Stachyose hydrate, 97+%
Stachyose hydrate, 97+%
Thermo Scientific Chemicals

Stachyose hydrate, 97+%

CAS: 54261-98-2 | C24H42O21 | 666.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 226080010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54261-98-2
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-({[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}methyl)oxane-3,4,5-triol
Molecular FormulaC24H42O21
InChI KeyUQZIYBXSHAGNOE-CYDMKDQUNA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.0 %
Water=<12 % (K.F.)
Specific optical rotation+129° to +140° (20°C, 589 nm) (c=4, water)
Solubility(5 % in water) Clear to slightly hazy colorless
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only