Aristolochic acid I, 96%
Aristolochic acid I, 96%
Aristolochic acid I, 96%
Thermo Scientific Chemicals

Aristolochic acid I, 96%

CAS: 313-67-7 | C17H11NO7 | 341.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number 226095000
Price (USD)
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS313-67-7
IUPAC Name8-methoxy-6-nitro-2H-phenanthro[3,4-d][1,3]dioxole-5-carboxylic acid
Molecular FormulaC17H11NO7
InChI KeyBBFQZRXNYIEMAW-UHFFFAOYSA-N
SMILESCOC1=C2C=C(C3=C(C=C4OCOC4=C3C2=CC=C1)C(O)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only