Nalgene™ Polycarbonate, Carboy with Sanitary Neck
Thermo Scientific™
Nalgene™ Polycarbonate, Carboy with Sanitary Neck
For Use With: Designed for use as receiver or dispensing vessel in biopharmaceutical applications; Autoclavable; Color: Clear
Have Questions?
Catalog NumberQuantity
2261-0050Case of 4
Catalog number 2261-0050
Price (USD)
1,764.00
Case of 4
Quantity:
Case of 4
Price (USD)
1,764.00
Case of 4
Clean the one-piece molded, non-threaded Thermo Scientific™ Nalgene™ Polycarbonate Sanitary Carboy more easily than threaded containers. Designed for use as receiver or dispensing vessel in biopharmaceutical applications, the 3 in. sanitary flange molded on the neck accepts standard flange fittings. A clamping closure system securely seals and will not back off.
  • Solid ported sanitary end caps are available
  • Made of same polycarbonate (PC) resin as Nalgene PC Clearboys and PC Bottles, permitting switch to Sanitary PC Carboys without material validation issues
  • To seal closure, use with Nalgene Heavy Duty Clamp (Cat. No. 2865-0300) and Gasket (Cat. No. 2672-0300)
  • Autoclavable
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)5.3 gal.
Closure Size3 in.
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
IncludesSanitary Neck
No. per Case4/Cs.
No. per Pack1 Ea.
QuantityCase of 4
Volume (English) Brim6.3 gal.
Volume (Metric) Brim24 L
Workflow StepUpstream media/buffer prep and storage. Upstream process solution storage
AutoclavableAutoclavable
Capacity (Metric)20 L
ColorClear
MaterialPolycarbonate
Product TypeCarboy with Sanitary Neck
SterilityNon-sterile
Unit SizeCase of 4