Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format - FAQs
Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format - FAQs
View additional product information for Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format - FAQs (22622)
21 product FAQs found
What is the difference between SMCC and Sulfo-SMCC?
SMCC is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane-stabilized spacer arm (8.3 angstroms). Sulfo-SMCC is a similar compound but it has a sulfonate group on the NHS ring which makes it water-soluble whereas SMCC is water-insoluble and needs to be dissolved first in an organic solvent like DMF or DMSO. Subsequent dilution into aqueous reaction buffer is generally possible, and most protein reactants will remain soluble if the final concentration of organic solvent is less than 10%.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Is the Thermo Scientific IP Lysis Buffer similar to RIPA buffer?
The Thermo Scientific IP Lysis Buffer (Cat. No. 87787) is a mammalian whole cell lysis buffer based on a modified RIPA buffer formulation without SDS. This moderate strength lysis buffer is able to effectively solubilize cellular proteins but does not liberate genomic DNA or disrupt protein complexes like RIPA buffer.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can Sulfo-SMCC crosslinker reagent be reconstituted in an organic solvent (DMF or DMSO) and added to an aqueous reaction mixture?
Yes, Sulfo-SMCC can be reconstituted in an organic solvent (DMF or DMSO) and added to an aqueous reaction mixture.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can the Sulfo-SMCC cross-linking agent be added directly to the reaction mixture?
Yes, Sulfo-SMCC can be added directly to the reaction mixture.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Is there a method to detect if a protein contains accessible sulfhydryl groups?
Yes, Ellman's reagent (Cat. No. 22582) can be utilized to determine if a protein contains free sulfhydryl groups.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
When crosslinking using Sulfo-SMCC, can proteins be modified to add free sulthydryls?
Yes, Trauts Reagent (Cat. No. 26101) will react with primary amine groups to generate a free sulfhydryl group. SATA (Cat No. 26102) will react with primary amine groups to generate a protected free sulfhydryl group that can be released prior to conjugation.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Will the maleimide chemistry in Sulfo-SMCC crosslinker react with disulfides?
No, the disulfides must first be reduced to free sulfhydryls utilizing a reducing agent. If the reducing agent contains free -SH (DTT, B-ME etc.) it must first be removed by desalting. Dialysis may not be used as a substitute for desalting, since over time, disulfide linkages will re-form. 10-50 mM EDTA in the reaction buffer can help slow the re-oxidation of sulfhydryls to disulfides.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How is the Sulfo-SMCC crosslinking reaction quenched?
1M Tris (or glycine), pH 7.5 can be added to the reaction mixture to a final concentration of approximately 20-50 mM to quench the NHS-Ester, or the product may be desalted to remove un-reacted cross-linker. Freshly prepared cysteine may be used to quench the maleimide chemistry.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can I make up a Sulfo-SMCC stock solution and store the cross-linker long-term?
Long-term storage is not recommended since the NHS-ester group is susceptible to hydrolysis. Hydrolysis begins upon reconstitution. Even organic solvents such as DMSO and DMF are hygroscopic, as a result, they tend to absorb water, which in turn also promotes the hydrolysis of the NHS-Ester.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What does "store under nitrogen" mean for Sulfo-SMCC crosslinker?
A gentle stream of nitrogen gas is held over the opening of the vial to flush the oxygen from the vial. The cap is then immediately placed on the vial. If storing under nitrogen, caution should be taken to ensure that the stream of nitrogen is gentle enough, so that the powdered product will not be blown out of the bottle. If nitrogen is not available, weigh the material as quickly as possible and immediately cap. Store desiccated.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the recommended storage condition for Sulfo-SMCC crosslinker?
The product ships under ambient temperature conditions. Upon receipt, the product should be stored at -20 degrees C in a dessicator or with a desiccant bag, and the vial should be warmed to room temperature before opening.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the expiration date for the Sulfo-SMCC crosslinker?
This product has no set expiration date. We monitor the bulk material in such a manner that allows us to warranty the product for one year from the date of shipment. It may still be usable beyond the one year if handled properly. However, using material older than one year is typically not recommended since the material easily hydrolyzes upon exposure to moisture.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How much Sulfo-SMCC cross-linking agent should I use?
A 10-20 fold molar excess of cross-linker to protein is recommended for protein solutions greater than 5 mg/mL. A 20-100 fold molar excess of cross-linker to protein is recommended for protein solutions less than 5 mg/mL.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the recommended concentration of the Sulfo-SMCC crosslinker stock solution?
An initial stock concentration of 5-12 mM (2 - 5 mg/mL) is recommended, or the cross-linking reagent may be added directly to the reaction mixture.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What buffers are not recommended for protein crosslinking using Sulfo-SMCC?
Buffers containing primary amines, primary sulfhydryls, Tris, ethanolamine, glycine, cysteine or lysine should not be used, since they will quench the reaction. Imidazole, glutathione, or sulfhydryl containing reducing reagents are also not recommended as buffer components.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the recommended sample buffer and reconstitution buffer to obtain optimal protein cross-linking using Sulfo-SMCC?
We recommend using 100 mM sodium phosphate, 0.15 M NaCl, pH 7.2 (PBS). HEPES, bicarbonate/carbonate, and borate buffers may be substituted for PBS. Other buffers may also be substituted provided they do not contain primary amines or primary sulfhydryls. Recommended pH is 7-9. The reaction can be performed over a pH range of 7-9, however as the pH increases, the rate of hydrolysis of the NHS-Ester also increases as well as the likelihood that the maleimide will react with amines. Once the cross-linker is reconstituted, it must be used immediately.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the difference between a homo- and a hetero- bifunctional cross-linker?
A homobifunctional cross-linker has the same reactive group on both ends, for example two amine-reactive groups. It is used in a one-step conjugation where the two molecules are mixed prior to the addition of the cross-linker. This leads to less efficient cross-linking because polymers can easily be created. A heterobifunctional cross-linker has two different reactive groups, for example an amine-reactive group on one end and a sulhydryl reactive group on the other. These can be used in two-step conjugations by reacting the fIrst end with the molecule containing its reactive group, and then adding the molecule that is reactive towards the other end. This leads to much more efficient cross-linking because polymerization does not occur.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.