Guanosine 5'-monophosphate, disodium salt hydrate, 97%
Guanosine 5'-monophosphate, disodium salt hydrate, 97%
Guanosine 5'-monophosphate, disodium salt hydrate, 97%
Thermo Scientific Chemicals

Guanosine 5'-monophosphate, disodium salt hydrate, 97%

CAS: 5550-12-9 | C10H12N5Na2O8P | 407.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 226600050
Price (EUR)
129,65
Online exclusive
144,00
Save 14,35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
129,65
Online exclusive
144,00
Save 14,35
Each
Chemical Identifiers
CAS5550-12-9
IUPAC Namedisodium [(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Molecular FormulaC10H12N5Na2O8P
InChI KeyPVBRXXAAPNGWGE-KHRSEZDTNA-L
SMILES[Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=96.0 % (On dry substance)
UV spectrophotometrymolar absorptivity 13700 +/- 4%, pH 7
Heavy metals (as Pb)=<10 ppm
UV spectrophotometry>=97.0 % (lambda max. 252 nm, pH 7)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only