Creatine monohydrate, 99%
Creatine monohydrate, 99%
Creatine monohydrate, 99%
Thermo Scientific Chemicals

Creatine monohydrate, 99%

Creatine monohydrate, CAS # 6020-87-7, is the monohydrate form of creatine, an amino acid found primarily in skeletal muscle cells that is important for rapid energy replenishment. | CAS: 6020-87-7 | C4H9N3O2·H2O | 149.15 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 226795000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS6020-87-7
IUPAC Name2-(N-methylcarbamimidamido)acetic acid hydrate
Molecular FormulaC4H11N3O3
InChI KeyMEJYXFHCRXAUIL-UHFFFAOYSA-N
SMILESO.CN(CC(O)=O)C(N)=N
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO498.5 to 102.0 %
HPLC>=98.5 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Creatine is an amino acid made naturally by the body that can also be acquired through dietary consumption
  • It is primarily stored in skeletal muscle tissue
  • Creatine is stored in a phosphate form, and upon depletion of ATP deposits, phosphocreatine is able to donate phosphate groups to rapidly replenish ATP
  • Creatine supplementation may be beneficial as a disease treatment, particularly in diseases of muscle wasting including cancer and chronic disease

Applications

  • Creatine may be used in the laboratory to study its potential for use as a treatment for metabolic and muscle wasting diseases
  • Creatine has been found to have antitumor, antiviral, and antidiabetic effects
  • It is also used in laboratory settings to study neurological disorders and has been found to be neuroprotective against hypoxic conditions
RUO – Research Use Only

General References:

  1. Turner, C.E; Byblow, W.D.; Gant, N. Creatine supplementation enhances corticomotor excitability and cognitive performance during oxygen deprivation. Journal of Neuroscience.2015, 35, (4), 1773-1780.
  2. Schuenke, M.D.; Brooks, N.D.; Hikida, R.S. Interactions of aging, overload, and creatine supplementation in rat plantaris muscle. Journal of Aging Research.2011, 2011, 393416.
  3. Wyss, M.; Kaddurah-Daouk, R. Creatine and creatinine metabolism. Physiol Rev. 2000 Jul;80(3):1107-213.