2-YT Broth
Invitrogen™

2-YT Broth

2-YT Broth is a nutrient-rich microbial growth medium used for cultivation of E. coli and growth of hosts for replicationRead more
Have Questions?
Catalog NumberQuantity
22712020500 g
Catalog number 22712020
Price (USD)
163.65
Online exclusive
174.00
Save 10.35
Each
Quantity:
500 g
Price (USD)
163.65
Online exclusive
174.00
Save 10.35
Each

2-YT Broth is a nutrient-rich microbial growth medium used for cultivation of E. coli and growth of hosts for replication of M13 vectors. The 2-YT formulation is similar to Lennox LB medium, but contains double the concentration of yeast extract and a 60% higher concentration of peptone. 2-YT medium allows cells to reach higher density and increase recombinant protein production compared to LB media formulations.

2-YT Broth formulation
Formulation per liter: 16 g SELECT Peptone 140 (pancreatic digest of casein), 10 g SELECT Yeast Extract, and 5 g NaCl.
Note: To prepare 1 L of 2-YT medium, weigh 31 g of powder and add distilled water to 1 L. Adjust the pH value to 7.0 with 5 N NaOH and sterilize by autoclaving for 20 minutes at 121°C.

Primary applications
• Growth of hosts for replication of M13 vectors
• Recombinant protein expression
• Achieve higher plasmid DNA yield

Find the media format that fits you needs
We offer a variety of ready-to-use media formulations including LB Broth, Terrific Broth and S.O.C Recovery Medium.
Explore our media formats for bacterial growth.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatBottle
Media Type2-YT Broth (Yeast Extract Tryptone)
Preparation MethodAutoclave
Quantity500 g
Shipping ConditionRoom Temperature
Product TypeDehydrated Culture Media
SpeciesEscherichia coli
Target Organism ClassColiforms
Unit SizeEach
Contents & Storage
• 2-YT Broth Powder (500 g)

Store at room temperature.