Citric acid, trisodium salt dihydrate, 99%, pure
Citric acid, trisodium salt dihydrate, 99%, pure
Citric acid, trisodium salt dihydrate, 99%, pure
Thermo Scientific Chemicals

Citric acid, trisodium salt dihydrate, 99%, pure

CAS: 1545801 | C6H9Na3O9 | 294.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
View all
Plastic Bottle
Catalog number 227135000
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.10
Save 6.45 (15%)
Each
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.10
Save 6.45 (15%)
Each
Chemical Identifiers
CAS6132-04-3
IUPAC Nametrisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Molecular FormulaC6H9Na3O9
InChI KeyNLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILESO.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 %
Water11 to 13 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only