1-Naphthyl acetate, 99%, Thermo Scientific Chemicals
1-Naphthyl acetate, 99%, Thermo Scientific Chemicals
1-Naphthyl acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Naphthyl acetate, 99%, Thermo Scientific Chemicals

CAS: 830-81-9 | Molecular Formula: C12H10O2 | Molecular Weight: 186.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
22725025025 gGlass bottle
Catalog number 227250250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS830-81-9
IUPAC Namenaphthalen-1-yl acetate
Molecular FormulaC12H10O2
InChI KeyVGKONPUVOVVNSU-UHFFFAOYSA-N
SMILESCC(=O)OC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light pink
Appearance (Form)Crystalline powder or crystals
Melting point42°C to 47°C
Infrared spectrumConforms
HPLC>=98.5 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only