Polyvinylpyrrolidone, cross-linked
Polyvinylpyrrolidone, cross-linked
Polyvinylpyrrolidone, cross-linked
Thermo Scientific Chemicals

Polyvinylpyrrolidone, cross-linked

CAS: 25249-54-1 | (C6H9NO)n | 111.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 227485000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS25249-54-1
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD00149016
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Loss on drying=<5 %
Heavy metals=<10 ppm
Sulfated ash=<0.1 %
pH5 to 8 (1 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only