4-Nitrophenyl sulfate, potassium salt, 99+%
4-Nitrophenyl sulfate, potassium salt, 99+%
4-Nitrophenyl sulfate, potassium salt, 99+%
Thermo Scientific Chemicals

4-Nitrophenyl sulfate, potassium salt, 99+%

CAS: 6217-68-1 | C6H4KNO6S | 257.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 227490010
Price (USD)
87.20
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
87.20
Each
Chemical Identifiers
CAS6217-68-1
SpecificationsSpecification SheetSpecification Sheet
HPLC=<0.05 % free p-nitrophenol
HPLC>=99.0 %
Infrared spectrumConforms
Appearance (Color)Yellow to khaki to brown
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only