Chloramine-T trihydrate, 97+%
Chloramine-T trihydrate, 97+%
Chloramine-T trihydrate, 97+%
Thermo Scientific Chemicals

Chloramine-T trihydrate, 97+%

Chloramine-T trihydrate, >-97%, C7H14ClNNaO5S, CAS Number-7080-50-4, 5kg, 97+%, White, 192 deg.C, 281.69, GHS Signal Word: Danger | CAS: 7080-50-4 | C7H13ClNNaO5S | 281.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic Drum
Catalog number 227850050
Price (USD)
729.00
Each
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
729.00
Each
Chemical Identifiers
CAS7080-50-4
IUPAC Namesodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
Molecular FormulaC7H13ClNNaO5S
InChI KeyRNROOTWAJBXJBD-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration Iodometric97.0 to 103.0 %
Appearance of solution(5% in water) Clear solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only