ART™ Barrier Low-Retention Pipette Tips
Thermo Scientific™
ART™ Barrier Low-Retention Pipette Tips
Eliminate sample retention with these specially designed pipette tips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ModelTip StyleFit for Purpose
2279-051000 μLART 1000Ergonomic, Low RetentionSterile and PCR Ready
2139-0510 μLART 10Low Retention, Micro PointSterile and PCR Ready
2139-05PK10 μLART 10Low Retention, Micro PointSterile and PCR Ready
2140-0510 μLART 10 REACHExtended Length, Low Retention, Micro PointSterile and PCR Ready
2140-05PK10 μLART 10 REACHExtended Length, Low Retention, Micro PointSterile and PCR Ready
2149P-0520 μLART 20PLow Retention, Micro PointSterile and PCR Ready
2149P-05PK20 μLART 20PLow Retention, Micro PointSterile and PCR Ready
2069-05200 μLART 200Low Retention, Micro PointSterile and PCR Ready
2069-05PK200 μLART 200Low Retention, Micro PointSterile and PCR Ready
2279-05PK1000 μLART 1000Ergonomic, Low RetentionSterile and PCR Ready
Catalog number 2279-05
Price (USD)
608.02
Online exclusive
808.00
Save 199.98
Case of 3200
Volume (Metric):
1000 μL
Model:
ART 1000
Tip Style:
Ergonomic, Low Retention
Fit for Purpose:
Sterile and PCR Ready
Price (USD)
608.02
Online exclusive
808.00
Save 199.98
Case of 3200
Get Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips, which are specifically designed for eliminating sample retention within the pipette tip.
  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Tips seal when exposed to potential contaminants
  • Certified RNase, DNase, ATP, and Endotoxin Free
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 1000, Filtered, Sterile, Rack
Length (English)3.13 in.
Length (Metric)7.9cm
ModelART 1000
PackagingRack, 8 x 100
AutoclavableNot Autoclavable
ColorClear
FilteredFiltered
Fit for PurposeSterile and PCR Ready
FormatRack
Product LineART
SterilitySterile
Tip StyleErgonomic, Low Retention
Volume (Metric)1000 μL
Unit SizeCase of 3200