Search
View additional product information for Pierce™ Power System Welcome Pack - FAQs (22830SPCL)
1 product FAQs found
The Thermo Scientific Power System (Cat. No. 22830) consists of the Thermo Scientific Power Station (Cat. No. 22838) with activated Staining and Blotting Software, the Thermo Scientific Power Stain Cassette (Cat. No. 22836), and the Thermo Scientific Power Blot Cassette (Cat. No. 22835). It is a combination of the Power Stainer and the Power Blotter.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.