Search
View additional product information for Pierce™ Power Blot Cassette - FAQs (22835)
6 product FAQs found
An alternative for the discontinued Power Blot Cassette (Cat. No. 22835) is the Power Blotter Cassette XL (Cat. No. PB0003). It functions in the same way but is a different color.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Any staining procedure that uses over 50% alcohol will dehydrate a gel. Simply rehydrate the gel in some water.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
No. Each cassette can only be used for its intended purpose; Power Blot Cassette for blotting (fast protein transfer) and Power Stain Cassette for protein staining. We have optimized the surface area to yield the best results for each application. The smaller stain cassette works better for staining while the larger blot cassette works better for blotting.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
No. It is not possible to insert the Power Blot Cassette and access the Power Staining protocol or vice versa.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Yes. The Thermo Scientific Power Station automatically recognizes the cassette that is inserted and initializes the correct software menu system.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
The Thermo Scientific Power System (Cat. No. 22830) consists of the Thermo Scientific Power Station (Cat. No. 22838) with activated Staining and Blotting Software, the Thermo Scientific Power Stain Cassette (Cat. No. 22836), and the Thermo Scientific Power Blot Cassette (Cat. No. 22835). It is a combination of the Power Stainer and the Power Blotter.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.