3-Isobutyl-1-methylxanthine, 99+%
3-Isobutyl-1-methylxanthine, 99+%
3-Isobutyl-1-methylxanthine, 99+%
Thermo Scientific Chemicals

3-Isobutyl-1-methylxanthine, 99+%

CAS: 28822-58-4 | C10H14N4O2 | 222.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 228420010
Price (USD)
194.65
Special offer
Online exclusive
Ends: 28-Dec-2026
229.00
Save 34.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
194.65
Special offer
Online exclusive
Ends: 28-Dec-2026
229.00
Save 34.35
Each
Chemical Identifiers
CAS28822-58-4
IUPAC Name1-methyl-3-(2-methylpropyl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC10H14N4O2
InChI KeyAPIXJSLKIYYUKG-UHFFFAOYSA-N
SMILESCC(C)CN1C2=C(NC=N2)C(=O)N(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Melting point200°C to 203°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only