Difco Casamino Acids, vitamin assay, is an animal origin (AO) acid hydrolysis of casein having low sodium chloride and ironRead more
Have Questions?
Change view
Catalog Number
Quantity
For Use With (Application)
Grade
228820
100 g
Vitamin assay
Vitamin Assay
228830
500 g
Vitamin assay
Vitamin Assay
2 Options
Catalog number 228820
Price (USD)
162.00
Each
Quantity:
100 g
For Use With (Application):
Vitamin assay
Grade:
Vitamin Assay
Price (USD)
162.00
Each
Difco Casamino Acids, vitamin assay, is an animal origin (AO) acid hydrolysis of casein having low sodium chloride and iron concentrations. It is an excellent supplement for media formulations where nitrogen requirements are minimal, including diphtheria toxin production.