Difco™ Casamino Acids, vitamin assay
Actual product may vary
Difco™ Casamino Acids, vitamin assay
Difco™ Casamino Acids, vitamin assay
Gibco™

Difco™ Casamino Acids, vitamin assay

Difco Casamino Acids, vitamin assay, is an animal origin (AO) acid hydrolysis of casein having low sodium chloride and ironRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFor Use With (Application)Grade
228830500 gVitamin assayVitamin Assay
228820100 gVitamin assayVitamin Assay
Catalog number 228830
Price (MXN)
-
Quantity:
500 g
For Use With (Application):
Vitamin assay
Grade:
Vitamin Assay
Difco Casamino Acids, vitamin assay, is an animal origin (AO) acid hydrolysis of casein having low sodium chloride and iron concentrations. It is an excellent supplement for media formulations where nitrogen requirements are minimal, including diphtheria toxin production.

Specifications
AdditiveCasamino Acid
Certifications/ComplianceAOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014
For Use With (Application)Vitamin assay
GradeVitamin Assay
Packaging TypeBottle
Product LineDifco
Quantity500 g
Shelf Life1825 Days
FormDehydrated Powder
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.