Patulin, 99%
Patulin, 99%
Patulin, 99%
Thermo Scientific Chemicals

Patulin, 99%

CAS: 149-29-1 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Packaging:
Glass bottle
Catalog number 228870050
Price (USD)
184.65
Special offer
Online exclusive
Ends: 28-Dec-2026
217.00
Save 32.35
Each
Quantity:
5 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
184.65
Special offer
Online exclusive
Ends: 28-Dec-2026
217.00
Save 32.35
Each
Chemical Identifiers
CAS149-29-1
IUPAC Name4-hydroxy-2H,4H,6H-furo[3,2-c]pyran-2-one
Molecular FormulaC7H6O4
InChI KeyZRWPUFFVAOMMNM-UHFFFAOYNA-N
SMILESOC1OCC=C2OC(=O)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
HPLC>=98.5 %
Melting point105°C to 114°C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only