3,3',5,5'-Tetramethylbenzidine, 99+%
3,3',5,5'-Tetramethylbenzidine, 99+%
3,3',5,5'-Tetramethylbenzidine, 99+%
Thermo Scientific Chemicals

3,3',5,5'-Tetramethylbenzidine, 99+%

CAS: 54827-17-7 | C16H20N2 | 240.35 g/mol
Have Questions?
Catalog NumberQuantityPackaging
22928025025 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 229280250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54827-17-7
IUPAC Name3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular FormulaC16H20N2
InChI KeyUAIUNKRWKOVEES-UHFFFAOYSA-N
SMILESCC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Melting point167°C to 172°C
Loss on drying=<0.5 %
Appearance (Form)Crystalline powder
Titration with HClO498.0 to 102.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only