Citrinin, 98%
Citrinin, 98%
Citrinin, 98%
Thermo Scientific Chemicals

Citrinin, 98%

CAS: 518-75-2 | C13H14O5 | 250.25 g/mol
Have Questions?
Catalog NumberQuantity
2295800505 mg
Catalog number 229580050
Price (USD)
-
Quantity:
5 mg
Request bulk or custom format
Chemical Identifiers
CAS518-75-2
IUPAC Name(3R,4S)-7-(dihydroxymethylidene)-3,4,5-trimethyl-4,6,7,8-tetrahydro-3H-2-benzopyran-6,8-dione
Molecular FormulaC13H14O5
InChI KeyJLWQFKMKUKQXMW-IYSWYEEDSA-N
SMILESC[C@H]1OC=C2C(=O)C(=C(O)O)C(=O)C(C)=C2[C@@H]1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only