Malachite Green oxalate, pure, high purity biological stain
Malachite Green oxalate, pure, high purity biological stain
Malachite Green oxalate, pure, high purity biological stain
Thermo Scientific Chemicals

Malachite Green oxalate, pure, high purity biological stain

Malachite Green oxalate, C52H54N4O12, CAS Number-2437-29-8, C.I. 42000, Basic Green 4, 500g, 0.9 to 1.15, 13, 745, 219-441-7, 927.03, Pure, Danger | CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 229781000
Price (USD)
170.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
170.00
Each
Chemical Identifiers
CAS2437-29-8
IUPAC Namebis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
Molecular FormulaC52H54N4O12
InChI KeyCNYGFPPAGUCRIC-UHFFFAOYSA-L
SMILESOC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
UV Lambda max615 to 619 nm (water)
Absorption Ratio P-15/P+150.9 to 1.15
Specific absorbance A (1%/1cm)>=1800 (at lambda max.)
Infrared spectrumConforms
Loss on drying=<10 % (105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only