Poroszyme® Immobilized Pepsin, Bulk Media

Catalog number: 2313200

Applied Biosystems™  Related applications: Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Bulk Immobilized Pepsin to POROS® media. The immobilized proteolytic enzyme provides complete, automated digestions in 1 - 5 minutes, with high sample recovery.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Analytical Column, Bulk Resin, Immobilized Enzyme Media
Flow Rate: 1000 to 5000 cm⁄hr
For Use With (Equipment): FPLC
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Immobilized Phase: Pepsin
Product Line: Applied Biosystems™, Poroszyme®
Product Size: 0.1 mL
Sample Type (Specific): Protein
Shipping Condition: Wet Ice
Support Matrix: Polystyrenedivinylbenzene

Contents & storage

Store this medium at 4°C.

Documents

Manuals & protocols