1-Butanol, 99.5%, for analysis
1-Butanol, 99.5%, for analysis
1-Butanol, 99.5%, for analysis
Thermo Scientific Chemicals

1-Butanol, 99.5%, for analysis

CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 232080025
Price (EUR)
111,00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
111,00
Each
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.1 % (Coulometric)
Substance colored by H2SO4Passes test
Barium (Ba)=<0.1 ppm
Cobalt (Co)=<0.02 ppm
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only