Pierce™ BCA Protein Assay Reagent A
Thermo Scientific™

Pierce™ BCA Protein Assay Reagent A

BCA Protein Assay Reagent A is a component of the Pierce BCA Protein Assay Kit, a two-component, high-precision, detergent-compatible assayRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
232231 L
232223.75 L
23221250 mL
23228500 mL
Catalog number 23223
Price (USD)
192.65
Online exclusive
219.00
Save 26.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
192.65
Online exclusive
219.00
Save 26.35
Each
BCA Protein Assay Reagent A is a component of the Pierce BCA Protein Assay Kit, a two-component, high-precision, detergent-compatible assay that is used for total protein concentration determination compared to a protein standard. Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research. The Pierce BCA assay can be used to assess yields in whole cell lysates, affinity-column fractions, purified proteins samples, as well as to monitor protein contamination in industrial applications. Compared to most dye-binding methods, the BCA assay is affected much less by protein compositional differences, providing greater concentration accuracy.

Compare all available BCA protein assays ›

Features of the Pierce BCA Protein Assay Kit (Reagents A and B) include:
• Colorimetric—estimate visually or measure with a standard spectrophotometer or plate reader at 562 nm
• Uniformity—exhibits less protein-to-protein variation than dye-binding methods (Bradford)
• Compatibility—unaffected by typical concentrations of most ionic and non-ionic detergents
• Assay time—30-min incubation; much easier and four times faster than classical Lowry methods
• Assay range—linear working range for BSA of 20 to 2000 μg/mL
• Sensitivity—detect down to 5 μg/mL with the enhanced protocol

How the assay works
The BCA Protein Assay combines the well-known reduction of Cu2+ to Cu1+ by protein in an alkaline medium with the highly sensitive and selective colorimetric detection of the cuprous cation (Cu1+) by bicinchoninic acid (BCA). The first step is the chelation of copper with protein in an alkaline environment to form a light blue complex. In this reaction, known as the biuret reaction, peptides containing three or more amino acid residues form a colored chelate complex with cupric ions in an alkaline environment containing sodium potassium tartrate.

In the second step of the color development reaction, BCA reacts with the reduced (cuprous) cation that was formed in step one. The intense purple-colored reaction product results from the chelation of two molecules of BCA with one cuprous ion. The BCA/copper complex is water soluble and exhibits a strong linear absorbance at 562 nm with increasing protein concentrations. The complex is approximately 100 times more sensitive (lower limit of detection) than the pale blue color of the first reaction.

The reaction is strongly influenced by four amino acid residues (cysteine, cystine, tyrosine, and tryptophan) in the amino acid sequence of the protein. However, unlike Coomassie dye-binding methods, the universal peptide backbone also contributes to color formation, helping to minimize variability caused by protein compositional differences.

Related products
Pierce BCA Protein Assay Kit
Pierce BCA Protein Assay Reagent B
Pierce BCA Solid
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayBCA Assay
DescriptionPierce BCA Protein Assay Reagent A
For Use With (Application)Solution-based Detection, Absorbance
For Use With (Equipment)Spectrophotometer, Microplate Reader
Product LinePierce
Product TypeProtein Quantitation Assay
Quantity1 L
SpecificityNot Target-Specific
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

Which Thermo Scientific protein assay is the best or the most reliable?

The choice of protein assay is dependent on preferences related to assay speed, accuracy and sensitivity, as well as interfering substances in the sample to be assayed. BCA has less protein-to-protein variation, is compatible with most detergents, and has larger working range. Pierce Bradford Plus Protein Assay Kit (Cat. No. 23236) is compatible with reducing sugars, is more sensitive and is faster and easier to use. For a comparison of different protein assays and compatible reagents, see our Tech Tip: Protein Quantitation Assay Compatibility Table (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.

What substances interfere with the BCA Protein Assay?

Reducing agents, copper chelators and solutions with very high buffering capacities will interfere with the BCA assay. Reducing agents reduce the copper and will produce a high background. Copper chelators bind the copper and prevent it from being detected by the BCA reagent. High-capacity buffers prevent the BCA from reaching its optimal alkaline pH. For a complete list of compatible substances, please refer to the BCA Protein Assay Kit product instructions.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.

What is the sample to working reagent ratio when using the BCA Protein Assay Kit?

For the Microplate Protocol the sample to working reagent (WR) ratio is 1:8 (25 µl sample plus 200 µl WR). If the sample amount is limited, a 1:20 ratio may be used (10 µl sample); in this case, however, the detection range of the assay will be limited to 125-2,000 µg/ml. For the Test Tube Procedure (Standard or Enhanced Protocols) the ratio is 1:20 (0.1 ml sample plus 2.0 ml WR). In the Micro BCA Assay Kit (for both Microplate and Test Tube Procedures), the ratio is 1:1 (1 part sample plus 1 part WR).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.

What is the detection range for the BCA Protein Assay?

For the standard protocol, the detection range is 20-2,000 µg/ml. For the enhanced test tube protocol, the detection range is 5-250 µg/ml. The Micro BCA Protein Assay Reagent will detect 0.5-20 µg/ml of protein in the test tube assay and 1-20 µg/ml for the microplate assay.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.

How can interfering substances be eliminated from the protein sample when using the BCA Protein Assay Kit?

There are several ways to adjust a sample to be compatible with the BCA Protein Assay: • Remove the interfering substance by dialysis or gel filtration. • If the starting protein concentration of the sample is high, dilute the sample to the point that the substance no longer interferes. • Eliminate interference by copper-chelating agents by increasing the amount of copper in the working reagent (use 4 ml or 6 ml of Reagent B/100 ml of Reagent A instead of the 2 ml of Reagent B/100 ml of Reagent A that is called for in the instructions). • Precipitate sample proteins with cold acetone or trichloroacetic acid (TCA).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.