1-Hydroxypyrene, 99+%
1-Hydroxypyrene, 99+%
1-Hydroxypyrene, 99+%
Thermo Scientific Chemicals

1-Hydroxypyrene, 99+%

CAS: 5315-79-7 | C16H10O | 218.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Catalog number 232270010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5315-79-7
IUPAC Namepyren-1-ol
Molecular FormulaC16H10O
InChI KeyBIJNHUAPTJVVNQ-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Beige to brown
HPLC>=99.0 %
Appearance (Form)Powder or needles
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only