Nalgene™ HDPE, Low-Profile Carboy - FAQs
View additional product information for Nalgene™ HDPE, Low-Profile Carboy - FAQs (2323-0008)
1 product FAQs found
Do you offer replacement carboy spigots?
The standard Nalgene polyproylene quick action spigot (blue handle and body, white threaded ring for connection to carboy) Cat. No. is 6422-0010 or 6422-0010PK. If the original carboy purchased was a solvent-resistant fluorinated carboy, the replacement (mostly white, with a blue cap on the handle) is the Nalgene Quick-Action Tefzel ETFE Spigot, Cat. No. 6432-0010.