Pierce™ Bradford Plus Protein Assay Kits
Thermo Scientific™

Pierce™ Bradford Plus Protein Assay Kits

The Pierce Bradford Plus Protein Assay Kit is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent used to quickly measure total protein concentration compared to a protein standard.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호농도
23236BSA: 2 mg/mL
A55866BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL
카탈로그 번호 23236
제품 가격(KRW)
333,000
온라인 행사
Ends: 31-Dec-2026
370,000
할인액 37,000
Each
농도:
BSA: 2 mg/mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
333,000
온라인 행사
Ends: 31-Dec-2026
370,000
할인액 37,000
Each

The Pierce Bradford Plus Protein Assay Kit is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent used to quickly measure total protein concentration compared to a protein standard. It provides increased linearity and half the protein-to-protein variability of other commercial Bradford assay formulations.

Kits are available with or without Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.

The Pierce Bradford Plus Protein Assay Kit is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent used to quickly measure total protein concentration compared to a protein standard. It provides increased linearity and half the protein-to-protein variability of other commercial Bradford assay formulations.

Kit options include Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.

Features of the Pierce Bradford Plus Protein Assay Kit include:

  • Colorimetric—measure with a standard spectrophotometer or plate reader at 595 nm
  • Easy to use—single reagent; no working reagent preparation required
  • Fast—almost immediate color development; add, mix, and read results
  • Wide assay range—detects protein concentration in the range 1 to 1500 μg/mL
  • Linear—improved linearity and response uniformity compared to traditional Bradford formulations
  • Flexible—microplate and cuvette protocols provided and adaptable to several target working ranges
  • Serial dilution–free—select kit options include Dilution-Free BSA Protein Standards that eliminate the need to perform serial dilutions when generating a standard curve

The assay is performed at room temperature. Simply add the sample to the tube or well containing reagent, and the resultant blue color is measured at 595 nm following a short room-temperature incubation. The protein assay is compatible with most salts, solvents, buffers, thiols, reducing substances, and metal-chelating agents encountered in protein samples. The assay can be performed in either test tube or microplate format.

How the Bradford Plus Assay detects protein

Use of Coomassie G-250 dye in a colorimetric reagent for the detection and quantitation of total protein was first described by Dr. Marion Bradford in 1976. In the acidic environment of the reagent, protein binds to the Coomassie dye. This results in a spectral shift from the reddish/brown form of the dye (absorbance maximum at 465 nm) to the blue form of the dye (absorbance maximum at 610 nm). The difference between the two forms of the dye is greatest at 595 nm, so that is the optimal wavelength to measure the blue color from the Coomassie dye-protein complex. If desired, the blue color can be measured at any wavelength between 575 nm and 615 nm. At the two extremes (575 nm and 615 nm) there is a loss of about 10% in the measured amount of color (absorbance) compared to that obtained at 595 nm.

Development of color in Coomassie dye-based (Bradford) protein assays has been associated with the presence of certain basic amino acids (primarily arginine, lysine, and histidine) in the protein. Van der Waals forces and hydrophobic interactions also participate in the binding of the dye by protein. The number of Coomassie dye ligands bound to each protein molecule is approximately proportional to the number of positive charges found on the protein. Free amino acids, peptides, and low molecular weight proteins do not produce color with Coomassie dye reagents. In general, the mass of a peptide or protein must be at least 3000 Daltons to be assayed with this reagent.

For Research Use Only.
사양
어세이Bradford Assay
농도BSA: 2 mg/mL
용도(애플리케이션)Solution-based Detection, Absorbance
용도(장비)Spectrophotometer, Microplate Reader
키트 구성품10 x 1 mL glass ampules
제품라인Pierce
제품 유형Protein Quantitation Assay
수량950 mL
특이성Not Target-Specific
충분630 Tube Assays or 3160 Microplate Assays
검출 방법Colorimetric
Unit SizeEach
구성 및 보관
Sufficient For: 630 tube assays or 3160 microplate assays
• Pierce Bradford Plus Assay Reagent, 950 mL
• Albumin Standard ampules, 2 mg/mL, 10 x 1 mL

Store Bradford Plus Assay Reagent at 4°C. Store unopened Albumin Standard ampules at room temperature.

자주 묻는 질문(FAQ)

Which Thermo Scientific protein assay is the best or the most reliable?

The choice of protein assay is dependent on preferences related to assay speed, accuracy and sensitivity, as well as interfering substances in the sample to be assayed. BCA has less protein-to-protein variation, is compatible with most detergents, and has larger working range. Pierce Bradford Plus Protein Assay Kit (Cat. No. 23236) is compatible with reducing sugars, is more sensitive and is faster and easier to use. For a comparison of different protein assays and compatible reagents, see our Tech Tip: Protein Quantitation Assay Compatibility Table (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0068-Protein-assay-compatibility.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.

What is the effect of temperature on the Pierce Bradford Plus Protein Assay Kit?

The absorbance readings at 595 nm obtained with the Pierce Bradford Plus Protein Assay Kit are dependent on the temperature of the reagent to some extent. As the reagent temperature increases to room temperature, the 595 nm readings will increase. Therefore, it is important that the Pierce Bradford Plus Protein Assay Reagent be at room temperature during the assay.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is the detection range of Pierce Bradford Plus Protein Assay Kit?

Pierce Bradford Plus Protein Assay Kit has a detection range of 1µg/ml to 1,500µg/ml of protein. The protein of interest must have a molecular weight greater than 3,000 daltons to be detected by the assay. For peptides smaller than 3,000 daltons, try using Fluoraldehyde Reagent Solution (Cat. No. 26025), which will fluorescently detect amino groups on small peptide.

Other peptide quantitation assays we offer are Thermo Scientific Pierce Quantitative Colorimetric Peptide Assay (Cat. No. 23275) and Thermo Scientific Pierce Quantitative Fluorometric Peptide Assay (Cat. No. 23290).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What are the advantages of the Pierce Bradford Plus Protein Assay Kit over the Bradford Method?

Pierce Bradford Plus Protein Assay Kit has several advantages over the Bradford Method:

- The standard curve is more linear.
- No reagent preparation is required.
- No dilution or filtering is necessary.
- The reaction is very fast (results in as little as 30 seconds).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What agents are compatible with Pierce Bradford Plus Protein Assay Kit?

The assay is compatible with many agents that are incompatible in other assays. It has been shown to be compatible with amine-containing buffers, reducing agents, chaotropic agents, organic solvents, sugars, antimicrobial agents, DNA, protease inhibitors, and low concentrations of metal chelators and metals.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

인용 및 참조 문헌 (10)

인용 및 참조 문헌
Abstract
Co-association of methotrexate and SPIONs into anti-CD64 antibody-conjugated PLGA nanoparticles for theranostic application.
Authors:Moura CC, Segundo MA, Neves Jd, Reis S, Sarmento B
Journal:
PubMed ID:25364249
Rheumatoid arthritis (RA) is an autoimmune disease with severe consequences for the quality of life of sufferers. Regrettably, the inflammatory process involved remains unclear, and finding successful therapies as well as new means for its early diagnosis have proved to be daunting tasks. As macrophages are strongly associated with RA ... More
Differences in sperm protein abundance and carbonylation level in bull ejaculates of low and high quality.
Authors:Mostek A, Westfalewicz B, Slowinska M, Dietrich MA, Judycka S, Ciereszko A
Journal:PLoS One
PubMed ID:30427859
'In breeding and insemination centres, significant variation in bull ejaculate quality is often observed between individuals and also within the same individual. Low-quality semen does not qualify for cryopreservation and is rejected, generating economic loss. The mechanisms underlying the formation of low-quality ejaculates are poorly understood; therefore, the aim of ... More
Mitogen-Activated Protein Kinase Cross-Talk Interaction Modulates the Production of Melanins in Aspergillus fumigatus.
Authors:Manfiolli AO, Siqueira FS, Dos Reis TF, Van Dijck P, Schrevens S, Hoefgen S, Föge M, Straßburger M, de Assis LJ, Heinekamp T, Rocha MC, Janevska S, Brakhage AA, Malavazi I, Goldman GH, Valiante V
Journal:MBio
PubMed ID:30914505
'The pathogenic fungus'
ICAM3-Fc Outperforms Receptor-Specific Antibodies Targeted Nanoparticles to Dendritic Cells for Cross-Presentation.
Authors:Cruz LJ, Tacken PJ, van der Schoot JMS, Rueda F, Torensma R, Figdor CG
Journal:Molecules
PubMed ID:31083610
'Optimal targeting of nanoparticles (NP) to dendritic cells (DCs) receptors to deliver cancer-specific antigens is key to the efficient induction of anti-tumour immune responses. Poly (lactic-co-glycolic acid) (PLGA) nanoparticles containing tètanus toxoid and gp100 melanoma-associated antigen, toll-like receptor adjuvants were targeted to the DC-SIGN receptor in DCs by specific humanized ... More
Per-Arnt-Sim Kinase (PASK) Deficiency Increases Cellular Respiration on a Standard Diet and Decreases Liver Triglyceride Accumulation on a Western High-Fat High-Sugar Diet.
Authors:Pape JA, Newey CR, Burrell HR, Workman A, Perry K, Bikman BT, Bridgewater LC, Grose JH
Journal:Nutrients
PubMed ID:30558306
Diabetes and the related disease metabolic syndrome are epidemic in the United States, in part due to a shift in diet and decrease in physical exercise. PAS kinase is a sensory protein kinase associated with many of the phenotypes of these diseases, including hepatic triglyceride accumulation and metabolic dysregulation in ... More