2,3-Dichlorobenzaldehyde, 99%
2,3-Dichlorobenzaldehyde, 99%
2,3-Dichlorobenzaldehyde, 99%
Thermo Scientific Chemicals

2,3-Dichlorobenzaldehyde, 99%

CAS: 6334-18-5 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
23267025025 gGlass bottle
Catalog number 232670250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6334-18-5
IUPAC Name2,3-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyLLMLNAVBOAMOEE-UHFFFAOYSA-N
SMILESClC1=CC=CC(C=O)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow
Melting point60°C to 64°C
Appearance (Form)Powder or crystals and/or chunks
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only