Nunc™ Sealing Tapes, Aluminum Acrylate, 384 well plates - FAQs
Nunc™ Sealing Tapes, Aluminum Acrylate, 384 well plates - FAQs
View additional product information for Nunc™ Seals, Tapes, and Foils - FAQs (236272, 236366, AB0718, 236370, 250050, 241205, 5701TS1, 232702, 276014, 236707, 3501, 249720, 236710, 232698, 235306, 232699, AB0580, 232701, 235307, 236366PK, 236703)
1 product FAQs found
Can I boil Nunc Sealing Tape?
Our Nunc Stealing Tape is thermostable from -20 degrees C to 100 degrees C.