Pierce™ Quantitative Peroxide Assay Kit (Lipid) - FAQs
View additional product information for Pierce™ Quantitative Peroxide Assay Kit (Lipid) - FAQs (23285)
1 product FAQs found
Do you offer any protein assays that are protein specific?
We offer several “specialty” protein assays including: Protease assay kits (Cat. Nos. 23263 and 23266), Glycoprotein Carbohydrate Estimation Kit (Cat. No. 23260), Phosphoprotein Phosphate Estimation Kit (Cat. No. 23270), Quantitative Peroxide assay kits (Cat. Nos. 23280 and 23285), and Easy-Titer Assay kits for IgG and IgM.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.