4-Benzoylpiperidine hydrochloride, 98%
4-Benzoylpiperidine hydrochloride, 98%
4-Benzoylpiperidine hydrochloride, 98%
Thermo Scientific Chemicals

4-Benzoylpiperidine hydrochloride, 98%

CAS: 25519-80-6 | C12H15NO·ClH | 225.72 g/mol
数量:
1 g
10 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 232860100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS25519-80-6
IUPAC Name4-benzoylpiperidin-1-ium
Molecular FormulaC12H16NO
InChI KeyDDRCHUGHUHZNKZ-UHFFFAOYSA-O
SMILESO=C(C1CC[NH2+]CC1)C1=CC=CC=C1
さらに表示
Appearance (Color)White to off-white
Titration with HClO4>=97.5 %
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only