Petroleum ether, extra pure, boiling range 100-140°C
Petroleum ether, extra pure, boiling range 100-140°C
Petroleum ether, extra pure, boiling range 100-140°C
Thermo Scientific Chemicals

Petroleum ether, extra pure, boiling range 100-140°C

CAS: 64742-49-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 L
25 L
Packaging:
View all
Metal Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 233020250
Price (USD)
-
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
SpecificationsSpecification SheetSpecification Sheet
Benzene=<50 ppm
Boiling point98°C to 140°C
Appearance (Form)Clear liquid
Infrared spectrumConforms
Density(15°C) 0.737 to 0.747 g/ml
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only