4-Epianhydrotetracycline hydrochloride, can be used as a secondary standard
4-Epianhydrotetracycline hydrochloride, can be used as a secondary standard
4-Epianhydrotetracycline hydrochloride, can be used as a secondary standard
Thermo Scientific Chemicals

4-Epianhydrotetracycline hydrochloride, can be used as a secondary standard

4-Epianhydrotetracycline hydrochloride, CAS # 4465-65-0, is an antibiotic derivative of tetracycline. It is an important standard for monitoring tetracycline stability and toxicity in laboratory studies.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
233111000100mgGlass bottle
233115000500mgGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 233111000
Price (MXN)
-
Quantity:
100mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4465-65-0
IUPAC Name4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide hydrochloride
Molecular FormulaC22H23ClN2O7
InChI KeySPFAOPCHYIJPHJ-UHFFFAOYNA-N
SMILESCl.CN(C)C1C2CC3=C(C(O)=C4C(O)=CC=CC4=C3C)C(=O)C2(O)C(O)=C(C(N)=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Purity>=80 % (as is, calculated)
Infrared spectrumConforms
Appearance (Color)Orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • 4-Epianhydrotetracycline hydrochloride is a secondary degradation product formed by epimerization of tetracycline and dehydration at the C6 position to aromatize the B ring
  • It has 250-fold higher toxicity than either tetracycline or doxycycline depending on the system under study

Applications

  • 4-Epianhydrotetracycline hydrochloride is less toxic to E. coli, and thus suitable for use with vectors or systems designed upon elements of the (Tn10) tet operon of E. coli
  • This compound is also used in veterinary medicine research and as a secondary standard for application in quality control in pharmaceutical laboratories
RUO – Research Use Only