Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Thermo Scientific Chemicals

Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
数量:
25 mL
100 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 233280250
価格(JPY)
-
数量:
25 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS111-30-8
IUPAC Namepentanedial
Molecular FormulaC5H8O2
InChI KeySXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILESO=CCCCC=O
さらに表示
Color scale=<100 APHA
Specific gravity(20/20°C) 1.000 to 1.133
Appearance (Form)Clear to slightly hazy solution
Assay25 to 27 % w/w
pH3.1 to 4.5 (at 25°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only