Silver lactate, 96%
Thermo Scientific Chemicals

Silver lactate, 96%

CAS: 15768-18- | C3H5O3Ag | 196.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
View all
Glass bottle
Catalog number 233330100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS15768-18-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Grey-brown
Titration Argentometric>=95 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only