o-Phenetidine, 99%
o-Phenetidine, 99%
o-Phenetidine, 99%
Thermo Scientific Chemicals

o-Phenetidine, 99%

CAS: 94-70-2 | C8H11NO | 137.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2334800505 mLGlass bottle
Catalog number 233480050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS94-70-2
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.5540 to 1.5560 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear light yellow to red to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only