4-Hydroxy-2(5H)-furanone, 95%
4-Hydroxy-2(5H)-furanone, 95%
4-Hydroxy-2(5H)-furanone, 95%
Thermo Scientific Chemicals

4-Hydroxy-2(5H)-furanone, 95%

CAS: 541-57-1 | C4H4O3 | 100.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 235260050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS541-57-1
IUPAC Name4-hydroxy-2,5-dihydrofuran-2-one
Molecular FormulaC4H4O3
InChI KeyJZQBAGOECGRTSA-UHFFFAOYSA-N
SMILESOC1=CC(=O)OC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=94 %
Appearance (Color)Light yellow to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only