Bacto™ Brain Heart Infusion
Gibco™

Bacto™ Brain Heart Infusion

Bacto Brain Heart Infusion is used in the cultivation of fastidious and non-fastidious microorganisms, including bacteria (streptococci, pneumococci, and meningococci),Read more
Have Questions?
Catalog NumberQuantity
23730010 kg
Catalog number 237300
Price (MXN)
-
Quantity:
10 kg
Bacto Brain Heart Infusion is used in the cultivation of fastidious and non-fastidious microorganisms, including bacteria (streptococci, pneumococci, and meningococci), yeasts, and molds. Bacto Brain Heart Infusion is a general-purpose medium used to cultivate aerobic and anaerobic bacteria from a variety of clinical and nonclinical materials.

Specifications
Certifications/ComplianceAOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA
ColorLight tan
For Use With (Application)Cultivation of fastidious and non-fastidious microorganisms, including aerobic and anaerobic bateria, yeasts and molds
FormulationCalf brain (infusion from 200g) 7.7g/L, Beef heart (infusion from 250g) 9.8g/L, Proteose Peptone 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L
Media TypeBrain Heart Infusion
Packaging TypePail
Product LineBacto
Quantity10 kg
Shelf Life1825 Days
FormDehydrated (free-flowing, homogenous)
Product TypeBrain Heart Infusion
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.