Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Thermo Scientific™

Nalgene™ LDPE Economy Wash Bottles

Prevent separation when squeezing these LDPE economy wash bottles, which feature a tight-fitting draw tube and closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2401-0125125 mL24 mmCase of 48
2401-0125PK125 mL24 mmPack of 6
2401-0250250 mL24 mmCase of 36
2401-0250PK
also known as 2401-0250
250 mL24 mmPack of 6
2401-0500500 mL28 mmCase of 24
2401-0500PK500 mL28 mmPack of 6
2401-10001000 mL38-430 mmCase of 12
2401-1000PK
also known as 2401-1000
1000 mL38-430 mmPack of 4
Catalog number 2401-0125
Price (USD)
240.30
Online exclusive
280.00
Save 39.70
Case of 48
Capacity (Metric):
125 mL
Closure Size:
24 mm
Unit Size:
Case of 48
Price (USD)
240.30
Online exclusive
280.00
Save 39.70
Case of 48

High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

Features:

  • Polypropylene screw closure/stem with PPCO draw tube
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

Specifications
Capacity4 oz., 125 mL
Capacity (English)4 oz.
Closure MaterialPolypropylene
Closure Size24 mm
Closure TypeScrew Cap
Diameter (Metric)50.3 mm
Height (English)5.8 in.
Height (Metric)147 mm
IncludesStem and draw tube
No. per Case48
No. per Pack6
Product TypeEconomy Wash Bottle
ShapeCylindrical
Through the Closure DispensingYes
Capacity (Metric)125 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialLDPE
MouthNarrow
Unit SizeCase of 48