Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories
Actual product may vary
Thermo Scientific™

Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories

Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
24072671Tube Dispensing Cassette5 Pk.
24070290Tubing Set1 Ea.
24070297Long Tubing Set1 Ea.
24072670Tube Dispensing Cassette1 Ea.
24072672Tube Dispensing Cassette10 Pk.
24072677Tube Dispensing Cassette, Long1 Ea.
Catalog number 24072671
Price (USD)
3,719.70
Online exclusive
4,174.00
Save 454.30
Each
Product Type:
Tube Dispensing Cassette
Quantity:
5 Pk.
Price (USD)
3,719.70
Online exclusive
4,174.00
Save 454.30
Each

Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.

Ordering Information:

Cat Nos 24070297 and 24070290 include tip combs.

Specifications
AutoclavableAutoclavable (50 times at 121°C, 1 bar, 20 min.)
Diameter (Metric)Tip: 0.5 mm; Tubing: 1.3 mm
For Use With (Equipment)Multidrop Combi, Multidrop 384, Multidrop DW
Quantity5 Pk.
Volume (Metric) Dead<7 mL
MaterialPolypropylene Tip, Silicone Tubing
Product TypeTube Dispensing Cassette
Volume (Metric)5 to 2500 μL
Unit SizeEach

Frequently asked questions (FAQs)

Can the SMART cassettes with RFID tags be used with the standard Multidrop Combi without SMART option or vice versa?

Yes, the cassettes are interchangeable, however, the calibration data cannot be viewed and the information about the cassette is not updated. Also standard cassettes can be used with the Multidrop Combi with SMART option.

What should I do if the liquid foams during dispensing with the Multidrop Combi?

Some liquids foam more easily than others. One option is to reduce the pump rotor speed. There are three dispensing speeds available: High, Med (medium), and Low.

What materials come into contact with liquids to be dispensed by the Multidrop Combi?

The Small tube plastic tip dispensing cassette, Cat. No. 24073290, and the Standard tube dispensing cassette, Cat. No. 24072670, are made of: PEEK, silicone, and PP (polypropylene).
The Small tube metal tip dispensing cassette, Cat. No. 24073295, is made of: PEEK, silicone, ruby, and stainless steel.

Do particles in the reagent affect dispensing with the Multidrop Combi?

Possibly. It is recommended that you ensure that there are no particles greater than 50 μm in the reagent to avoid blockage of the tips, especially when working with small tube cassettes.

What is the tip inner diameter (ID) of the dispensing cassettes of the Multidrop Combi?

The tip inner diameter (ID) is either 0.22 mm (Small tube plastic tip dispensing cassette, Cat. No. 24073290, and Small tube metal tip dispensing cassette, Cat. No. 24073295), or 0.5 mm (Standard tube dispensing cassette, Cat. No. 24072670).