Multidrop™ Combi SMART Reagent Dispenser Accessories
Thermo Scientific™

Multidrop™ Combi SMART Reagent Dispenser Accessories

Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Quantity
24073292SMART Small Plastic Tip0.5 to 50 μL1 Ea.
24073001SMART Small Plastic Tip0.5 to 50 μL5 Pk.
24072678SMART Long Standard5 to 2500 μL1 Ea.
24072676SMART Standard5 to 2500 μL5 Pk.
N22704SMART+ Standard5 to 2500 μL1 Ea.
N22706SMART+ Long Standard5 to 2500 μL1 Ea.
Catalog number 24073292
Price (USD)
1,122.00
Each
Description:
SMART Small Plastic Tip
Volume (Metric):
0.5 to 50 μL
Quantity:
1 Ea.
Price (USD)
1,122.00
Each

Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories.

  • Designed for use with Multidrop Combi SMART+ dispensers
  • Equipped with radio frequency identification (RFID) tags to store lifetime dispensing data and display remaining usage time
  • Available in standard and small tube versions
  • Distinguished from normal cassettes by antenna tags
  • Cat. No. N15133 to N15139 are to be used with the Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
  • Cat. No. 24073292, 24073001, 24073297, 24073002, 24072675, 24072678 and 24072676 are to be used with the Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM
Specifications
AutoclavableAutoclavable (10 times at 121°C, 1 bar, 20 min.)
DescriptionSMART Small Plastic Tip
Diameter (Metric)Tip: 0.4 mm; Tubing: 0.22 mm
For Use With (Equipment)Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM
Quantity1 Ea.
Volume (Metric) Dead<1 mL
MaterialPolypropylene Tip, Silicone Tubing
Product TypeTube Dispensing Cassette
Volume (Metric)0.5 to 50 μL
Unit SizeEach

Frequently asked questions (FAQs)

Can the SMART cassettes with RFID tags be used with the standard Multidrop Combi without SMART option or vice versa?

Yes, the cassettes are interchangeable, however, the calibration data cannot be viewed and the information about the cassette is not updated. Also standard cassettes can be used with the Multidrop Combi with SMART option.

What should I do if the liquid foams during dispensing with the Multidrop Combi?

Some liquids foam more easily than others. One option is to reduce the pump rotor speed. There are three dispensing speeds available: High, Med (medium), and Low.

What materials come into contact with liquids to be dispensed by the Multidrop Combi?

The Small tube plastic tip dispensing cassette, Cat. No. 24073290, and the Standard tube dispensing cassette, Cat. No. 24072670, are made of: PEEK, silicone, and PP (polypropylene).
The Small tube metal tip dispensing cassette, Cat. No. 24073295, is made of: PEEK, silicone, ruby, and stainless steel.

Do particles in the reagent affect dispensing with the Multidrop Combi?

Possibly. It is recommended that you ensure that there are no particles greater than 50 μm in the reagent to avoid blockage of the tips, especially when working with small tube cassettes.

What is the tip inner diameter (ID) of the dispensing cassettes of the Multidrop Combi?

The tip inner diameter (ID) is either 0.22 mm (Small tube plastic tip dispensing cassette, Cat. No. 24073290, and Small tube metal tip dispensing cassette, Cat. No. 24073295), or 0.5 mm (Standard tube dispensing cassette, Cat. No. 24072670).