KingFisher™ Flex 24 Deep Well heating block
Thermo Scientific™

KingFisher™ Flex 24 Deep Well heating block

The KingFisher™ Flex 24 Deep-Well Heating Block is compatible with the KingFisher™ Flex 24 Deep-Well Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 24 Deep-Well Head.
Have Questions?
Catalog NumberQuantity
24075440Each
Catalog number 24075440
Price (USD)
710.00
Each
Quantity:
Each
Price (USD)
710.00
Each

The KingFisher™ Flex 24 Deep-Well Heating Block is compatible with the KingFisher™ Flex 24 Deep-Well Plate when used on the KingFisher™ Flex Magnetic Particle Processor with 24 Deep-Well Head.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)KingFisher™ Flex
Product LineKingFisher
QuantityEach
Shipping ConditionRoom Temperature
Capacity24 Samples
Format24 Deep-well Heating Block
Unit SizeEach

Frequently asked questions (FAQs)

Can the KingFisher Flex heating blocks be purchased separately?

Yes, here are the catalog numbers for standalone heating blocks:

- Cat. No. 24075420: KingFisher Flex 96 KF Heating Block
- Cat. No. 24075430: KingFisher Flex 96 Deep-Well Heating Block
- Cat. No. 24075440: KingFisher Flex 24 Deep-Well Heating Block

Does the KingFisher Flex Magnetic Particle Processor with 24 Deep-Well Head come with a heating block?

Yes, it comes with the KingFisher Flex 24 Deep-Well Heating Block.

I see that the KingFisher 24 Deep Well Head has a processing volume of 5 mL. Does that mean that I can process up to 5 mL of my sample?

No, the processing volume refers to the maximum volume that can be held in the 24 Deep Well plates. This incorporates the volume of binding buffers and beads, so when scaling up reaction volumes, it is important to make sure that the total volume does not exceed 5 mL.

My KingFisher Flex run was stopped due to an error message. Can I start the protocol from where it stopped?

No, once a protocol was stopped, it will restart from the beginning. The extraction procedure can either be finished manually by using a magnetic stand, or a modified version of the original KingFisher Flex script would have to be created that starts off at the point where the run was stopped.

I have a 12-plate protocol but there are only 8 slots on the KingFisher Flex instrument turntable. What should I do with the remaining 4 plates?

Set up the protocol on the BindIt software. After the first 8 plates are done, the instrument will pause and instruct you to switch out the plates.