Nunc™ Square BioAssay Dishes
Thermo Scientific™

Nunc™ Square BioAssay Dishes

Screen large numbers of colonies for cloning experiments, culturing bacteria, fungi and agar diffusion assays using Thermo Scientific™ Nunc™ Squre BioAssay dishes.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Surface Treatment
24083525 mmUntreated
16650825 mmNunclon Delta
24084520 mmUntreated
Catalog number 240835
Price (USD)
282.65
Online exclusive
307.00
Save 24.35
Case of 16
Height (Metric):
25 mm
Surface Treatment:
Untreated
Price (USD)
282.65
Online exclusive
307.00
Save 24.35
Case of 16

Available in cell culture-treated and non-treated surfaces, the versatile Thermo Scientific™ Nunc™ Square BioAssay dishes are useful for screening large numbers of colonies, culturing bacteria and fungi, or providing a contained environment for other culture vessels.

Standard Height Dish:
  • External dimensions: 245 x 245 x 25mm
  • Lid height: 9.8mm
  • Available in non-treated or Nunclon Delta cell culture-treated surface
  • Optically clear and sterile

Low Profile Dish:

  • External dimensions: 241 x 241 x 20mm
  • Lid height: 10.3mm
  • Available in non-treated surface
  • Fits robotic instruments for genomic screening and colony picking
  • Optically clear and sterile
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Height (English)25 in.
No. per Case16
No. per Pack4
ShapeSquare
TypeMulti-Well Plate
Volume (Metric) Working225 mL
Bottom ShapeFlat
Capacity (Metric)225 mL
Culture Area500 sq. cm
Height (Metric)25 mm
SterilitySterile
Surface TreatmentUntreated
Unit SizeCase of 16

Frequently asked questions (FAQs)

Do you offer a Nunc square petri dish with a grid on the bottom?

We offer Nunc Square BioAssay Dishes, but this product does not have a grid.