Sesame oil
Thermo Scientific Chemicals

Sesame oil

CAS: 8008-74-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 241000010
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Chemical Identifiers
CAS8008-74-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Acid value=<0.6 mg KOH/g
Appearance (Form)Oily liquid
Refractive index1.4700 to 1.4760 (20°C, 589 nm)
Peroxide value=<10 meq/kg
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only