Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2
Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2
Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2
Acros Organics

Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2

CAS: 124-43-6 | CH6N2O3 | 94.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 241020051
also known as 24102
HSN 28470000
Price (INR)
83,790.00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (INR)
83,790.00
Each
Chemical Identifiers
CAS124-43-6
IUPAC Nameperoxol; urea
Molecular FormulaCH6N2O3
InChI KeyAQLJVWUFPCUVLO-UHFFFAOYSA-N
SMILESOO.NC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with KMnO4>=33.5 % (H2O2)
Stabilizer1 % Sodium pyrophosphate
Appearance (Form)Tablets
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only